missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyoxyethylene bis(amine), M.W. 1,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as PEG modifier, its main products include high-qualified intermediates, various of amino acid and APIs. This Polymer used in the preparation of enzyme conjugates soluble in organic solvents. Promising drug carrier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Granular Solid |
Solubility Information | It is soluble in organic solvents. It is insoluble in water. |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Chemical Name or Material | Polyoxyethylene bis (amine), M.W. 1,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'