Learn More
Short List Polynuclear Aromatic Hydrocarbons Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 8270-AF-M
Additional Details : Weight : 0.01000kg ??product.attributeSet.Transport?? : UN number : 1593 Chem class : 6.1 Pack group : III
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Acenaphthene | 83-32-9 | 2000μg/mL |
Acenaphthylene | 208-96-8 | 2000μg/mL |
Anthracene | 120-12-7 | 2000μg/mL |
Benz(a)anthracene | 56-55-3 | 2000μg/mL |
Benzo(a)pyrene | 50-32-8 | 2000μg/mL |
Benzo(b)fluoranthene | 205-99-2 | 2000μg/mL |
Benzo(g,h,i)perylene | 191-24-2 | 2000μg/mL |
Benzo(k)fluoranthene | 207-08-9 | 2000μg/mL |
Chrysene | 218-01-9 | 2000μg/mL |
Dibenz(a,h)anthracene | 53-70-3 | 2000μg/mL |
Fluoranthene | 206-44-0 | 2000μg/mL |
Fluorene | 86-73-7 | 2000μg/mL |
Indeno(1,2,3-c,d)pyrene | 193-39-5 | 2000μg/mL |
Naphthalene | 91-20-3 | 2000μg/mL |
Phenanthrene | 85-01-8 | 2000μg/mL |
Pyrene | 129-00-0 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Polynuclear Aromatic Hydrocarbons Mix | |
2mL amber prescored glass ampule | |
MeCl | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
SW-846 Method 8270C | |
For US EPA Method 8270C | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Safety and Handling
- Polynuclear Aromatic Hydrocarbons Mix
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P363-Wash contaminated clothing before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dichloromethane, benzo[a]pyrene, dibenz[a,h]anthracene, benz[a]anthracene, phenanthrene pure, fluore
Product intended for laboratory use only