Learn More
Polyhexamethylenbiguanid hydrochloride 100 μg/mL in Water, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Polyhexamethylenebiguanide hydrochloride |
| CAS | 32289-58-0 |
| Analyte or Component Names | Polyhexamethylenebiguanide hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | (C8 H17 N5)n . Cl H |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Poly[iminocarbonimidoyliminocarbonimidoylimino-1,6-hexanediyl] hydrochloride, Poly(iminoimidocarbonyliminoimidocarbonyliminohexamethylene) hydrochloride, Acticide SR 1296, Arlagard E, BG 1, BG-IR, Biobac ZS, Biodoctor, Cosmocil, Cosmocil CQ, Cosmocil PG, Lavasept, Lonzabac BG, Lonzabac BG 1, Marukacide AV, PHMB, PP 073, Polihexanide, Poly(hexamethylenebiguanide) hydrochloride, Poly(hexamethylenediguanide) hydrochloride, Polyhexanide, Prontosan, Proxel IB, Reputex 20, Vantocil 100, Vantocil IB, Vantocil P, Vantocil TG, Vantosan |
| Solvent or Matrix | Water |
| Molecular Weight (g/mol) | 219.72 |
| Show More |