Learn More
Macherey-Nagel™ Polygram CEL 300-50 UV254 Glass Plates
Brand:  Macherey-Nagel™ 808063
Additional Details : Weight : 0.01000kg
Product Code. 10194851
  • 308.00 € / Pack of 20
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
Specifications
20 Pk.
UV254
20 x 20 cm
Glass Plate
0.5 mm