Learn More
Macherey-Nagel™ Polygram CEL 300-50 Glass Plates
Brand:  Macherey-Nagel™ 808053
Additional Details : Weight : 1.00000kg
Product Code. 10206911
  • 308.00 € / Pack of 20
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
Specifications
20 Pk.
Glass Plate
0.5 mm
20 x 20 cm