Learn More
Polyethylene glycol dimethacrylate, M.W. 3,400, Thermo Scientific Chemicals
Description
Polyethylene glycol dimethacrylate is used as adhesives, coatings, sealants, photo resists, solder masks and photopolymers. Polyethylene Glycol Dimethacrylate forms homopolymers and copolymers. Copolymers of Polyethylene Glycol Dimethacrylate can be prepared with acrylic acid and its salts, amides and esters, and with methacrylates, acrylonitrile, maleic acid esters, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene, unsaturated polyesters and drying oils. It is also used as a very useful feedstock for chemical syntheses. It is employed as a cross linker for PVC plastisols. Polyethylene Glycol 200 Dimethacrylate is incorporated in cosmetic encapsulation resins and dental polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Granular Solid |
Molecular Formula | CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3 |
Solubility Information | It is soluble in water. |
Quantity | 1 g |
Sensitivity | Light and moisture sensitive |
Chemical Name or Material | Polyethylene glycol dimethacrylate, M.W. 3,400 |
RUO – Research Use Only