missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 95.5-96.5% hydrolyzed, M.W. approx. 85,000-124,000, Thermo Scientific Chemicals

Catalog Number p-7077186
24.73 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
1 kg
2.5 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
9002-89-5
(C2H4O)n
44.05
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
11199
ethenol
OC(-*)C-*
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
9002-89-5
(C2H4O)n
44.05
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
11199
ethenol
OC(-*)C-*

Indicator

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 230.0°C to 240.0°C
pH 4.5 to 6.5 (4% aq. soln.)
Infrared Spectrum Authentic
Linear Formula [-CH2CH(OH)-]n
Quantity 100 g
Merck Index 15,7706
Solubility Information Solubility in water: soluble in hot water.
Viscosity 27-31 mPa.s (20°C 4% aq. sol.)
Percent Purity 95.5-96.5%
Chemical Name or Material Poly(vinyl alcohol)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'