missing translation for 'onlineSavingsMsg'
Learn More

Platinum AAS Standard, 1000 mg/L Pt in HCl10%, Honeywell Fluka™

Brand:  Honeywell Fluka E4058-100ML

Additional Details : Weight : 0.10300kg ??product.attributeSet.Transport?? : UN number : 1789 Chem class : 8 Pack group : III

 View more versions of this product

Product Code. 16654364

  • 181.97 € / 100mL

Estimated Shipment: 24-01-2022
Add to basket

Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

Specifications

Specifications

Platinum AAS Standard
1.03 g/cm3
100 mL
Standards and Modifiers for AAS
<2
Videos
SDS
Documents
Certificates
Promotions

Promotions