missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on activated carbon powder, Type B105022-5, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Catalog Number 15424035
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7440-6-6
MFCD00011179
This item is not returnable. View return policy
7440-6-6
MFCD00011179

Platinum, 5% on activated carbon powder, Type B105022-5 is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones, and aromatics. It is also used for hydrogenolysis of cyclopropanes. Further, it is involved in the oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray to Black
Odor Odorless
Quantity 5 g
Solubility Information Insoluble in water.
Physical Form Powder
Chemical Name or Material Platinum, 5% on activated carbon powder, Type B105022-5, Standard, Reduced, Nominally 50% water wet

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'