Learn More
Welch™ PK 2 DC Rotary Vacuum Pump
Brand: Welch™ 322001
Additional Details : Weight : 0.10000kg
Specifications
30/36 L/min. | |
165 mm | |
120 w | |
50/60 Hz | |
330 mm | |
<1 x 10-2 mbar |
230 V 50/60 Hz | |
PK 2 DC | |
8 kg | |
170 mm | |
Rotary Vane Vacuum Pump | |
230 V |