Learn More
ROBU VitraPOR™ Pipeline Filter
Pipeline Filter with tube adapters for connection.
83.15 € - 131.00 €
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Sterility | Non-sterile |
Includes | Tube Adapters |
Product Code | Brand | Porosity | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Diameter (Metric) | Price | Quantity & Availability | ||||
10022141
|
ROBU VitraPOR™
23602 |
2 | 60 mm |
94.20 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
16863046
|
ROBU VitraPOR™
23300 |
0 | 30 mm |
83.15 €
Pack of 2 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
16883046
|
ROBU VitraPOR™
23302 |
2 | 30 mm |
83.15 €
Pack of 2 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10441802
|
ROBU VitraPOR™
23603 |
3 | 60 mm |
94.20 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
10460262
|
ROBU VitraPOR™
23604 |
4 | 60 mm |
101.00 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
10632982
|
ROBU VitraPOR™
23605 |
5 | 60 mm |
130.00 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
16893046
|
ROBU VitraPOR™
23303 |
3 | 30 mm |
83.15 €
Pack of 2 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
16813056
|
ROBU VitraPOR™
23305 |
5 | 30 mm |
131.00 €
Pack of 2 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10683752
|
ROBU VitraPOR™
23601 |
1 | 60 mm |
94.20 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
10746082
|
ROBU VitraPOR™
23600 |
0 | 60 mm |
94.20 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
16873046
|
ROBU VitraPOR™
23301 |
1 | 30 mm |
83.15 €
Pack of 2 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
16803056
|
ROBU VitraPOR™
23304 |
4 | 30 mm |
90.70 €
Pack of 2 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Non-sterile | |
Tube Adapters |