missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PIP5K2A (Human) Recombinant Protein
Human PIP5K2A full-length ORF ( AAH18034, 1 a.a. - 406 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | AAH18034 |
---|---|
Gene ID (Entrez) | 5305 |
Name | phosphatidylinositol-5-phosphate 4-kinase, type II, alpha |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139121
|
Abnova™
H00005305-P01.10UG |
10 μg |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149121
|
Abnova™
H00005305-P01.25UG |
25 μg |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MATPGNLGSSVLASKTKTKKKHFVAQKVKLFRASDPLLSVLMWGVNHSINELSHVQIPVMLMPDDFKAYSKIKVDNHLFNKENMPSHFKFKEYCPMVFRNLRERFGIDDQDFQNSLTRSAPLPNDSQARSGARFHTSYDKRYIIKTITSEDVAEMHNILKKYHQYIVECHGITLLPQFLGMYRLNVDGVEIYVIVTRNVFSHRLSVYRKYDLKGSTVAREASDKEKAKELPTLKDNDFINEGQKIYIDDNNKKVFLEKLKKDVEFLAQLKLMDYSLLVGIHDVERAEQEEVECEENDGEEEGESDGTHPVGTPPDSPGNTLNSSPPLAPGEFDPNIDVYGIKCHENSPRKEVYFMAIIDILTHYDAKKKAAHAAKTVKHGAGAEISTVNPEQYSKRFLDFIGHILT
Specifications
AAH18034 | |
phosphatidylinositol-5-phosphate 4-kinase, type II, alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
FLJ13267, PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5305 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PIP4K2A | |
GST |