missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PI3 (Human) Recombinant Protein
Human PI3 full-length ORF ( AAH10952, 1 a.a. - 117 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | AAH10952 |
---|---|
Gene ID (Entrez) | 5266 |
Name | peptidase inhibitor 3, skin-derived |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139061
|
Abnova™
H00005266-P01.10UG |
10 μg |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149061
|
Abnova™
H00005266-P01.25UG |
25 μg |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MRASSFLIVVVFLIAGTLVLEAAVTGVPVKGQDTVKGRVPFNGQDPVKGQVSVKGQDKVKAQEPVKGPVSTKPGSCPIILIRCAMLNPPNRCLKDTDCPGIKKCCEGSCGMACFVPQ
Specifications
AAH10952 | |
peptidase inhibitor 3, skin-derived | |
125% SDS-PAGE Stained with Coomassie Blue | |
ESI, MGC13613, SKALP, WAP3, WFDC14, cementoin | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5266 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PI3 | |
GST |