missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TGFBR1 (Ser165) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA540298
Additional Details : Weight : 0.00910kg
Description
This antibody is predicted to react with human and rat based on sequence homology.
Phospho-TGFBR1 (Ser165) Polyclonal antibody specifically detects Phospho-TGFBR1 (Ser165) in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.Specifications
Phospho-TGFBR1 (Ser165) | |
Polyclonal | |
Unconjugated | |
TGFBR1 | |
AAT5; activin A receptor type II-like kinase, 53kDa; activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ACVRLK4; ALK5; Alk-5; AU017191; ESK2; ESS1; LDS1; LDS1A; LDS2A; MSSE; mutant transforming growth factor beta receptor I; ser; serine/threonine-protein kinase receptor R4; SKR4; TbetaRI; tbetaR-I; tgf beta receptor 1; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFBR1; TGFR1; TGFR-1; Transfor; transforming growth factor beta receptor 1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; transforming growth factor, beta receptor I; transforming growth factor, beta receptor I (activin A receptor type II-like kinase, 53kDa); transforming growth factor-beta receptor type I | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21812, 29591, 7046 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P36897, P80204, Q64729 | |
TGFBR1 | |
Synthesized peptide derived from human TGFbeta RI around the phosphorylation site of S165. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |