missing translation for 'onlineSavingsMsg'
Learn More

Phospho-STAT3 (Tyr705) Mouse anti-Human, Mouse, APC, Clone: LUVNKLA, eBioscience™

Mouse Monoclonal Antibody

Marke:  Affymetrix eBioscience 17-9033-41

Weitere Details : Gewicht : 0.23750kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15528226

  • 124.00 € / 25 Tests


Auf Lager: 1
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Applications Reported: This LUVNKLA antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This LUVNKLA antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: We recommend using Protocol C: Two-step protocol: Fixation/Methanol. Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins and Protocol B: One-step protocol: intracellular (nuclear) proteins cannot be used. All Protocols can be found in the Flow Cytometry Protocols: "Staining Intracellular Antigens for Flow Cytometry Protocol" located in the Best Protocols Section under the Resources tab online. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. STAT3 belongs to the family of STAT (signal transducers and activators of transcription) proteins which are phosphorylated by receptor associated kinases, translocate to the nucleus, and act as transcription factors in response to cytokines and growth factors. Coactivators such as CREB-binding protein are required for the transcriptional activation by STAT3. STAT3 can also be activated by Interferon-alpha, Interferon-gamma, EGF, PDGF and IL6. Phosphorylation on tyrosine 705 by JAK1 and JAK2 is essential for STAT3 dimer formation, nuclear translocation, and DNA binding activity. In humans, the STAT3 gene is located on the q arm of chromosome 17. STAT3 has been shown to be activated by IFN-alpha but not IFN-beta. The transcription factors associated with STAT3 are c-Jun and cyclic AMP-responsive enhancer binding protein (CREB). STAT3 mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of STAT3 while the PIAS3 protein is a specific inhibitor of STAT3. Three alternatively spliced transcript variants encoding distinct isoforms of STAT3 have been described. Deletion of the STAT3 gene in knock-out mice was lethal at the early embryonic stage.
Spezifikation

Spezifikation

Phospho-STAT3 (Tyr705)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P40763, P42227
Signal Transducers and Activators of Transcription 3, Acute-phase response factor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
LUVNKLA
APC
STAT3
Liquid
STAT3
IgG2b, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
20848, 6774
Videos
Dokumentation

For Research Use Only.