Learn More
Macherey-Nagel™ Phosphatesmo KM Qualitative Test Paper
Brand: Macherey-Nagel™ 90607
Additional Details : Weight : 0.50000kg
Specifications
White to Violet | |
For detection of Sperm, acid phosphatase | |
25 Pack | |
Qualitative Test Paper |