Learn More
Pholcodine Monohydrate, Mikromol
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Pholcodine Monohydrate, Mikromol
Specifications
Specifications
| Chemical Name or Material | Pholcodine Monohydrate |
| CAS | 6254-99-5 |
| Analyte or Component Names | Pholcodine Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H30 N2 O4 . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Pholcodine, 7,8-Didehydro-4,5a-epoxy-17-methyl-3-[2-(morpholin-4-yl)ethoxy]morphinan-6a-ol monohydrate |
| InChI Formula | InChI=1S/C23H30N2O4.H2O/c1-24-7-6-23-16-3-4-18(26)22(23)29-21-19(5-2-15(20(21)23)14-17(16)24)28-13-10-25-8-11-27-12-9-25;/h2-5,16-18,22,26H,6-14H2,1H3;1H2/t16-,17+,18-,22-,23-;/m0./s1 |
| Solution Type | Neat |
| SMILES | O.CN1CC[C@]23[C@H]4Oc5c(OCCN6CCOCC6)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35 |
| Show More |