Learn More
Macherey Nagel Phenolphthalein Test Paper, Refill
pH indicator paper for phenolphthalein, without color scale, refill.
Brand:  Macherey Nagel 90713
Additional Details : Weight : 0.10000kg
Product Code. 10756532
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Simplest acid / base determination
    • Qualitative pH papers without color chart are simple, completely impregnated indicator papers.
    • They indicate if the pH of a solution is above or below the transition point (color change) and are useful to distinguish between acids and bases.
Specifications
Specifications
White to Red/pH 8.3 to 10.0
8.3 to 10
5 m
7 mm