missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative, Multi-Compendial, J.T. Baker™
864.50 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Specifications
Flash Point | 79.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 500 mL |
Specific Gravity | 1L = 1.06kg |
Formula Weight | 94.11 |
Grade | Multi-Compendial |
Chemical Name or Material | Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H400-Very toxic to aquatic life.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'