missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution, pH 12, Carl Roth™
Click to view available options
Quantity:
500 mL
1 L
Description
Buffer solution
pH 12.00
±0.05 (20 °C)
Ready to use
Glycine/Caustic soda lye/Sodium chloride
Specifications
Melting Point | 0°C |
Color | Colorless |
pH | 12 |
Physical Form | Liquid |
Quantity | 500 mL |
Solution Type | pH Buffer |
Vapor Pressure | 23hPa |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'