missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Penicillin V potassium salt, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.
Solubility
Soluble in water, dimethyl sulfoxide, and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.
Solubility
Soluble in water, dimethyl sulfoxide, and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
Specifications
Type | Penicillin V Potassium Salt |
Physical Form | Powder |
Melting Point | 197°C to 202°C |
Beilstein | 3899451 |
Merck Index | 14,7100 |
Solubility Information | Soluble in water,dimethyl sulfoxide,and methanol. |
Formula Weight | 388.48 |
Color | White |
Quantity | 10 g |
Chemical Name or Material | Penicillin V potassium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'