Learn More
Bode Chemie™ Peha-Soft™ Powder-Free Glove
Click to view available options
Size:
X-Small
Small
Large
X-Large
Description
- Disposable examination gloves made of soft latex, powder-free
- Easy to put on thanks to the inner coating
- Textured finger area for optimal grip
- Highly elastic, tear-resistant and safe
- Excellent fit and excellent tactile sensitivity
- To wear left and right.
Specifications
Certifications/Compliance | Approved for Contact with Food |
Color | White |
Disposable | Single-use |
Latex Free | No |
Material | Latex |
Powdered | Powder Free |
Product Type | Examination Gloves |
Size | X-Small |