missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD273 (B7-DC) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0464
Additional Details : Weight : 0.01000kg
Description
PDL-2 Synthetic Peptide, PEP-0464, from Invitrogen.
This peptide corresponds to 16 amino acids near the center of human PDL-2PEP-0464 can be used as a blocking peptide with polyclonal antibody PA5-20344.Specifications
80380 | |
purified | |
Human CD273 (B7-DC) | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9BQ51 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
PDCD1LG2 | |
0.02% sodium azide | |
50 μg | |
PBS |