Learn More
Greiner Bio-One PCR Strip Tubes without Cap
For use in research. Greiner Bio-One PCR Strip Tubes without Cap are free of detectable DNase, Rnase and human DNA.
Brand:  Greiner Bio-One 673273
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 10073361
Quantity Price
1 184.00 € / Pack of 125
10 156.31 € / Pack of 125
Estimated Shipment: 07-12-2021
Add to basket
Description
Description
  • Non-Pyrogenic
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
Specifications
Specifications
Polypropylene
PCR Tube Strip
0.2 mL
Non-sterile
Red
PCR