missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Parathyroid Hormone (3-34), bovine

172.00 € - 288.00 €

Specifications

CAS 51257-86-4
Content And Storage Store at -20°C
Format Solid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428269
SDS View Documents Promotion Details
Thermo Scientific™
J66777.!D
0.5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15438269
SDS View Documents Promotion Details
Thermo Scientific™
J66777.M^
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

51257-86-4
Store at -20°C
Solid
Videos
SDS
Documents

Documents

Product Certifications