missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ PACAP (6-27), Human, Ovine, Rat

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66872.M¬

Additional Details : CAS : 136134-68-4 Weight : 0.00001kg

 View more versions of this product

Product Code. 15418409

  • 241.00 € / 1mg


Estimated Shipment: 16-07-2021
Add to basket

Description

Description

Specifications

Specifications

2638.13
Human, Ovine, Rat
Solid
RUO
PACAP (6-27), human, ovine, rat
HPLC
1mg
Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2
-30° to -10°C
Solid
Videos
SDS
Documents
Certificates

For Research Use Only