missing translation for 'onlineSavingsMsg'
Learn More

Oxamyl, 95%, Thermo Scientific™

Catalog Number 15565186
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
23135-22-0
C7H13N3O3S
219.26
KZAUOCCYDRDERY-UITAMQMPSA-N
(Z)-[(dimethylcarbamoyl)(methylsulfanyl)methylidene]amino N-methylcarbamate
CNC(=O)O\N=C(/SC)C(=O)N(C)C
This item is not returnable. View return policy
23135-22-0
C7H13N3O3S
219.26
KZAUOCCYDRDERY-UITAMQMPSA-N
(Z)-[(dimethylcarbamoyl)(methylsulfanyl)methylidene]amino N-methylcarbamate
CNC(=O)O\N=C(/SC)C(=O)N(C)C
Quantity 1 g
Assay Percent Range 95%
Formula Weight 219.26
Chemical Name or Material Oxamyl
compliance-icons compliance-icons
Product Identifier
  • Oxamyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'