missing translation for 'onlineSavingsMsg'
Learn More

Orteronel, MedChemExpress

Product Code. p-200100360
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470367

Brand: Medchem Express HY10505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Orteronel (TAK-700) is a highly selective inhibitor of human 17,20-lyase (CYP17) with IC50 of 38 nM, and exhibits >1000-fold selectivity over other CYPs such as 11-hydroxylase and CYP3A4.

Orteronel (TAK-700) is a highly selective inhibitor of human 17,20-lyase (CYP17) with IC50 of 38 nM, and exhibits >1000-fold selectivity over other CYPs such as 11-hydroxylase and CYP3A4.

Specifications

Chemical Name or Material Orteronel
CAS 566939-85-3
Color White
Purity Grade Notes Research
Molecular Formula C18H17N3O2
Quantity 50 mg
Synonym TAK-700
Solubility Information DMSO : 14.29 mg/mL (46.49 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2C=C([C@@]3(O)CCN4C=NC=C43)C=CC2=C1)NC
Molecular Weight (g/mol) 307.35
Formula Weight 307.35
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.