Learn More
Kolmi Hopen™ OP-R™ Type IIR Splash-resistant Surgical Mask with Earloops
This splash-resistant surgical mask is complete with an anti-fog nose piece.
Brand: Kolmi Hopen™ M36201-30
4.51 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Resistant to human liquid projection up to 120 mmHg
- Soft polypropylene with non-woven edges
- Anatomical folding reduces exposure to face
- Integrated anti-fog nose piece
- Fiberglass-free filter
- Production in ISO 8 clean room (particle and microbiological control)
Specifications
Green | |
Polypropylene | |
Pleated | |
Single-use | |
Yes |
Earloops | |
175 x 90 mm | |
(EU) 2017/745, ISO 13485, EN 14683, EN ISO 14971 | |
Medical | |
50 Pk. |