Learn More
OLFML2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554343
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PTSIPATTTT ATTTPTPTTS LLPTEPPSGP EVSSQGREAS CEGTLRAVDP PVRHHSYGRH EGAWMKDPAA RDDRIYVTNY YY Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.
OLFML2A Polyclonal antibody specifically detects OLFML2A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
OLFML2A | |
Polyclonal | |
Unconjugated | |
OLFML2A | |
4932431K08Rik; mFLJ00237; olfactomedin like 2A; olfactomedin-like 2A; olfactomedin-like protein 2A; OLFML2A; Photomedin-1; PRO34319; UNQ9394/PRO34319 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
169611 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68BL7 | |
OLFML2A | |
Recombinant protein corresponding to Human OLFML2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only