Learn More
Ohaus™ OIML F2 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F2 Tolerances
Brand:  Ohaus™ 30390841
Additional Details : Weight : 2.11000kg
 View more versions of this product
Product Code. 16652212
  • 1050.00 € / Each
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
Specifications
NVLAP Accredited Certificate
1 kg to 1 g
13/Pk.
Troemner Alloy 8
Protective Case
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers