Learn More
Octadecanoic-12-d1 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Octadecanoic-12-D Acid |
| CAS | 62163-41-1 |
| Molecular Formula | C18 D H35 O2 |
| Quantity | 0.25 g |
| Synonym | Octadecanoic-12-d acid (9 CI), Stearic acid-12-d, Stearic-12-d acid, 12-Monodeuterostearic acid, [12-d]-Stearic acid, Octadecanoic-12-D acid |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i7D |
| SMILES | [2 H]C(CCCCCC)CCCCCCCCCCC(=O)O |
| IUPAC Name | 12-deuteriooctadecanoic acid |
| Molecular Weight (g/mol) | 285.483 |
| Formula Weight | 285.278 g/mol |
| Show More |