missing translation for 'onlineSavingsMsg'
Learn More

(+)-O,O'-Dipivaloyl-D-tartaric acid, 98+%, Thermo Scientific™

Catalog Number p-7037968
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
76769-55-6
C14H22O8
318.32
MFCD00015634
UFHJEZDFEHUYCR-YZYOREDDNA-N
16212279
CC(C)(C)C(=O)O[C@@H]([C@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O
This item is not returnable. View return policy
76769-55-6
C14H22O8
318.32
MFCD00015634
UFHJEZDFEHUYCR-YZYOREDDNA-N
16212279
CC(C)(C)C(=O)O[C@@H]([C@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 124°C to 126°C
Quantity 1 g
Beilstein 6894779
Sensitivity Hygroscopic
Synonym +-dipivaloyl-d-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, d-+-dipivaloyl tartaric acid, pubchem8173, --bis 2,2-dimethylpropionyl-l-tartaric acid, dipivaloyl-l-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, ?-o,o'-di-pivaloyl-l-tartaric acid, +-o,o'-di-pivaloyl-d-tartaric acid
Optical Rotation +24° (c=1.7 in Dioxane)
IUPAC Name (2S,3S)-2,3-bis[(2,2-dimethylpropanoyl)oxy]butanedioic acid
Formula Weight 318.32
Percent Purity ≥98%
Chemical Name or Material (+)-O,O'-Dipivaloyl-D-tartaric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'