missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals O-Acetyl-L-serine hydrochloride
Click to view available options
Quantity:
1 g
5 g
Description
O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.
Solubility
Soluble in ethanol almost transparency.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.
Solubility
Soluble in ethanol almost transparency.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Specifications
Color | White |
Solubility Information | Soluble in ethanol almost transparency. |
Formula Weight | 183.59 |
Quantity | 1 g |
Physical Form | Powder |
Chemical Name or Material | O-Acetyl-L-serine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'