Learn More
Thermo Scientific™ Nunc™ Barcode Scanners, USB
Track samples simply using these bar code scanners. Plug and Play function can automatically switch to read codes: 128A, 128B, 128C, 39 and Interleve 2/5.
Brand:  Thermo Scientific™ 330050
423.35 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.29250kg
Product Code. 10740295
  • 635.00 € / Each
Estimated Shipment: 23-04-2024
to see stock.
Add to basket
Description
Description
Track samples simply using Thermo Scientific™ Nunc™ Barcode Scanners. “Plug and Play” function can automatically switch to read codes: 128A, 128B, 128C, 39 and Interleve 2/5. Convenient and easy to use with an Excel spread sheet.
Durable, low cost, and easy to use–simply plug into a USB or keyboard port.
  • Scanners automatically switch between Code 128, Code 39, Interleave 2 of 5 and several other barcodes
  • Must be in contact with the barcode to read