missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ 8%, Tris-Glycine, Protein Gel, 26-well

Separation Range: 40 to 200 kd; Separation Type: Denaturing, Native; For Use With: Midi Gel Tank, XCell4 SureLock Midi

Brand:  Invitrogen™ WT0083BOX

Additional Details : Weight : 0.10000kg

Product Code. 10442873

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Includes: One box of 10 gels

Description

Description

  • Provide reproducible separation of a wide range of proteins into well-resolved bands
  • Individually packaged gels for optimal experiment design
  • Compatible with most protein standards; Usable for native and denaturing protein assays
  • Do not contain SDS and can be used to run proteins in native or in denatured form

Sample Loading Volume: 15μL; 1D Well

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Specifications

Specifications

XCell4 SureLock Midi-Cell
Tris-Glycine
Protein
Denaturing, Native
1 Box
8 cm
8%
Homogeneous Page
40 to 200 kDa
26-well
1D Well
13 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.