missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NLRX1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1132
Additional Details : Weight : 0.01000kg
Description
NOD5 Synthetic Peptide, PEP-1132, from Invitrogen.
This peptide corresponds to 14 amino acids near the amino terminus of human NOD5.PEP-1132 can be used as a blocking peptide with polyclonal antibody PA5-21018.Specifications
79671 | |
purified | |
Human NLRX1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q86UT6 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
NLRX1 | |
0.02% sodium azide | |
50 μg | |
PBS |