Learn More
Hanna Instruments™ Nitrite Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Nitrite Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured reagents for use with compatible benchtop and/or portable Hanna meters.
Brand:  Hanna Instruments™ HI9370703
Additional Details : Weight : 0.50000kg
Product Code. 10060881
  • 279.00 € / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of nitrite. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Reagent sets are available for low-range (using the EPA Diazotization method 354.1) and high-range (using the ferrous sulfate method) determinations
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Specifications
Nitrite low range determination
Individual packets
1 Set
EPA Diazotization method 354.1
Compatible Hanna benchtop and portable (HI96707) photometers
300
0.000 to 0.600mg/L nitrite (with HI96707) and 0.00 to 1.15mg/L nitrite (with benchtop photometers)
Water Testing Kit