Learn More
Hanna Instruments™ Nitrite Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Nitrite Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured reagents for use with compatible benchtop and/or portable Hanna meters.
Brand: Hanna Instruments™ HI9370703
Additional Details : Weight : 0.50000kg
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of nitrite. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Reagent sets are available for low-range (using the EPA Diazotization method 354.1) and high-range (using the ferrous sulfate method) determinations
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Nitrite low range determination | |
Individual packets | |
1 Set | |
EPA Diazotization method 354.1 |
Compatible Hanna benchtop and portable (HI96707) photometers | |
300 | |
0.000 to 0.600mg/L nitrite (with HI96707) and 0.00 to 1.15mg/L nitrite (with benchtop photometers) | |
Water Testing Kit |