Learn More
Nicotinyl Tartrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Nicotinyl Tartrate |
| CAS | 6164-87-0 |
| Analyte or Component Names | Nicotinyl Tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H7 N O . C4 H6 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Pyridinemethanol, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt) (9CI), 3-Pyridinemethanol, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), 3-Pyridinemethanol, tartrate (1:1) (salt) (8CI), 3-(Hydroxymethyl)pyridine tartrate, 3-Hydroxymethylpyridine hydrogen tartrate, 3-Pyridylcarbinol bitartrate, 3-Pyridylmethanol tartrate, Danaden, NSC 147492, Neu-2121, Niacol, Nicotinyl alcohol tartrate, Nicotinyl tartrate, Niltuvin, Pyridyl-3-carbinol tartrate, Radecol, Roniacol tartrate, Ronicol, Ronicol Retard, Ronicol Timespan, Ronicol tartrate, β-Pyridylcarbinol tartrate |
| InChI Formula | InChI=1S/C6H7NO.C4H6O6/c8-5-6-2-1-3-7-4-6;5-1(3(7)8)2(6)4(9)10/h1-4,8H,5H2;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1 |
| Solution Type | Neat |
| SMILES | OCc1cccnc1.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Show More |