Learn More
Nickel Copper foil, alloy 400, 0.51mm (0.02in) thick, Thermo Scientific Chemicals
118.00 € - 306.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15416785
|
Thermo Scientific Alfa Aesar
045148.GT |
150 x 150 mm |
118.00 €
Each |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15426785
|
Thermo Scientific Alfa Aesar
045148.HB |
300 x 300 mm |
306.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nickel Copper foil, alloy 400 finds application in marine engineering, chemical and hydro-carbon processings, heat exchangers, valves and pumps. Large use of it is made in the alkylation units, namely in the reacting section in contact with concentrated hydrofluoric acid.