missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NFE2L2 (Human) Recombinant Protein

Human NFE2L2 partial ORF (NP_006155.2, 351 a.a. - 450 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004780-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16074445

  • 518.16 € / 25µg

Estimated Shipment: 18-01-2022
Add to basket

Description

Description

  • Sequence: SPSVASPEHSVESSSYGDTLLGLSDSEVEELDSAPGSVKQNGPKTPVHSSGDMVQPLSPSQGQSTHVHDAQCENTPEKELPVSPGHRKTPFTKDKHSSRL
Specifications

Specifications

NP_006155.2
nuclear factor (erythroid-derived 2)-like 2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NFE2L2
GST
4780
Wheat germ expression system
25 μg
NRF2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Promotions

Promotions