missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Neuropeptide Y, Porcine

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66933.|D

Additional Details : CAS : 83589-17-7 Weight : 0.00001kg

 View more versions of this product

Product Code. 15418499

  • 201.00 € / 0.50mg


Estimated Shipment: 16-07-2021
Add to basket

Description

Description

Specifications

Specifications

4253.70
Porcine
Powder
RUO
Neuropeptide Y, porcine
HPLC
0.5mg
NPY
-30° to -10°C
Powder
Videos
SDS
Documents
Certificates

For Research Use Only