missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nepsilon-(2-chlorobenzyloxycarbonyl)-D-lysine, 98%, Thermo Scientific™

Catalog Number p-7054685
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
57096-11-4
C19H27ClN2O6
414.88
MFCD00065585
ATUMDPHEFWGCJF-GGYSOQFKNA-N
boc-d-lys 2-cl-z-oh, boc-n'-2-chloro-cbz-d-lysine, boc-d-lys 2-cl-cbz-oh, ambotzbaa1036, boc-d-lys 2-chloro-z-oh, boc-l-lys 2-cl-cbz-oh, n, a-boc-n, a-2-chloro-z-d-lysine, boc-d-lys 2-cl-z-oh tlc, n-boc-n'-2-chlorobenzyloxy carbonyl-d-lysine, n-tert-butoxy carbonyl-n'-2-chlorobenzyloxycarbonyl-d-lysine
7269325
CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1Cl)C(O)=O
This item is not returnable. View return policy
57096-11-4
C19H27ClN2O6
414.88
MFCD00065585
ATUMDPHEFWGCJF-GGYSOQFKNA-N
boc-d-lys 2-cl-z-oh, boc-n'-2-chloro-cbz-d-lysine, boc-d-lys 2-cl-cbz-oh, ambotzbaa1036, boc-d-lys 2-chloro-z-oh, boc-l-lys 2-cl-cbz-oh, n, a-boc-n, a-2-chloro-z-d-lysine, boc-d-lys 2-cl-z-oh tlc, n-boc-n'-2-chlorobenzyloxy carbonyl-d-lysine, n-tert-butoxy carbonyl-n'-2-chlorobenzyloxycarbonyl-d-lysine
7269325
CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1Cl)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Beilstein 5913148
IUPAC Name (2R)-6-[(2-chlorophenyl)methoxycarbonylamino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
Formula Weight 414.88
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nalpha-Boc-Nepsilon-(2-chlorobenzyloxycarbonyl)-D-lysine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'