missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-L-ornithine, 95%

Brand:  Alfa Aesar™ H62014.14

Additional Details : CAS : 21887-64-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15418618

  • 369.00 € / 25g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

21887-64-9
232.28
AMPVNPYPOOQUJF-ZETCQYMHSA-N
7018784
CC(C)(C)OC(=O)NC(CCCN)C(=O)O
C10H20N2O4
MFCD00076970
boc-orn-oh, boc-l-ornithine, n, a-boc-l-ornithine, l-ornithine, n2-1,1-dimethylethoxy carbonyl, 2s-5-amino-2-tert-butoxycarbonyl amino pentanoic acid, n2-boc-l-ornithine, nalpha-boc-l-ornithine, pubchem18983, nalpha-boc-l-ornithine;
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid

Specifications

Nalpha-Boc-L-ornithine
C10H20N2O4
4313353
AMPVNPYPOOQUJF-ZETCQYMHSA-N
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
7018784
Powder
White
25g
21887-64-9
MFCD00076970
boc-orn-oh, boc-l-ornithine, n, a-boc-l-ornithine, l-ornithine, n2-1,1-dimethylethoxy carbonyl, 2s-5-amino-2-tert-butoxycarbonyl amino pentanoic acid, n2-boc-l-ornithine, nalpha-boc-l-ornithine, pubchem18983, nalpha-boc-l-ornithine;
CC(C)(C)OC(=O)NC(CCCN)C(=O)O
232.28
232.28
95%
207°C to 208°C
Videos
SDS
Documents
Certificates