missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nafcillin sodium salt

Catalog Number p-7077884
Click to view available options
Quantity:
1 g
5 g
985-16-0
C21H21N2NaO5S
436.46
MFCD00056863
AYAPZOUDXCDGIF-FRFVDRIFSA-M
4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylicacid, 6-2-ethoxy-1-naphthalenyl carbonyl amino-3,3-dimethyl-7-oxo-, sodiumsalt 1:1 , 2s,5r,6r
131675018
(2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
[Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O
985-16-0
C21H21N2NaO5S
436.46
MFCD00056863
AYAPZOUDXCDGIF-FRFVDRIFSA-M
4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylicacid, 6-2-ethoxy-1-naphthalenyl carbonyl amino-3,3-dimethyl-7-oxo-, sodiumsalt 1:1 , 2s,5r,6r
131675018
(2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
[Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >181.0°C
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 436.46
Chemical Name or Material Nafcillin sodium salt
compliance-icons
Product Identifier
  • Nafcillin sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'