missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ N1,N11-Diethylnorspermine tetrahydrochloride
GREENER_CHOICE

Product Code. 18074682
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074682

Brand: Tocris Bioscience™ 0468/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Spermine and spermidine acetyltransferase potentiator

Highly potent inducer of spermidine/spermine N1-acetyltransferase, able to increase enzyme activity 200-1000 fold.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156886-85-0
Molecular Formula C13H36Cl4N4

Specifications

Quantity 50 mg
Formula Weight 390.27
Purity >98%
Molecular Weight (g/mol) 390.259
Chemical Name or Material N1, N11-Diethylnorspermine tetrahydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.