missing translation for 'onlineSavingsMsg'
Learn More

n-Octane-d{18}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7051970
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
17252-77-6
C8H18
132.342
MFCD00037626
TVMXDCGIABBOFY-VAZJTQEUSA-N
octane-d18, 2h18 octane, n-octane-d18, 2 h?? octane, octane-d18, 98 atom % d, n-octane-d isotopic 1g
519375
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecadeuteriooctane
CCCCCCCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
17252-77-6
C8H18
132.342
MFCD00037626
TVMXDCGIABBOFY-VAZJTQEUSA-N
octane-d18, 2h18 octane, n-octane-d18, 2 h?? octane, octane-d18, 98 atom % d, n-octane-d isotopic 1g
519375
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecadeuteriooctane
CCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 0.815 g/mL
Flash Point 15.6°C (60°F)
Quantity 1 g
Melting Point -57°C
Boiling Point 126°C
UN Number UN1262
Formula Weight 132.34
Percent Purity 99%
Chemical Name or Material n-Octane-d{18}, Isotopic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'