Learn More
N-Formylmethionine Specific Transfer Ribonucleic From E. coli MRE 600, MP Biomedicals™
Catalog Number p-7139885
Click to view available options
Quantity:
10 U
50 U
This item is not returnable. View return policy
63231-63-0
MFCD00132196
Ribonucleic acid
This item is not returnable. View return policy
63231-63-0
MFCD00132196
Ribonucleic acid
N-Formylmethionine Specific Transfer Ribonucleic From E. coli MRE 600 Activity=1300 ± 50 picomoles per A260 units, White Powder
  • Ribonucleic acid, transfer, N-formylmethionine-specific may be used as a substrate to study the specifity and kinetics of natural, mutated and chemically engineered N-formylmethionine-specific E. coli aminoacyl-tRNA synthetase(s) and methionyl-tRNA formyltransferase(s) (MTF).
  • Each T-RNA molecule is specific for and binds with a particular amino acid; more than one exists for each amino acid.
  • T-RNA performs three functions during protein synthesis: binds with its specific amino acid, recognizes the corresponding codon on mRNA and places the amino acid in the correct position for attachment to the polypeptide chain being formed & binds the polypeptide to the ribosome.
Color White
Quantity 10 U
Physical Form Powder