missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(4-thiazolyl)-L-alanine, 95%, Thermo Scientific Chemicals

Catalog Number 15418597
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
205528-32-1
C21H18N2O4S
394.445
MFCD00672568
LSBAZFASKHLHKB-IBGZPJMESA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiazol-4-yl propanoic acid, fmoc-l-4-thiazolyl-alanine, fmoc-beta-4-thiazolyl-ala-oh, n-fmoc-3-4-thiazolyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1,3-thiazol-4-yl propanoic acid, fmoc-3-4-thiazoyl-l-alanine, fmoc-4-thi-oh, fmoc-3-4-thiazolyl-alanine, fmoc-, a-4-thiazolyl-ala-oh, fmoc-3-4-thiazolyl-l-alanine
7020858
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1,3-thiazol-4-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=N4)C(=O)O
This item is not returnable. View return policy
205528-32-1
C21H18N2O4S
394.445
MFCD00672568
LSBAZFASKHLHKB-IBGZPJMESA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiazol-4-yl propanoic acid, fmoc-l-4-thiazolyl-alanine, fmoc-beta-4-thiazolyl-ala-oh, n-fmoc-3-4-thiazolyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1,3-thiazol-4-yl propanoic acid, fmoc-3-4-thiazoyl-l-alanine, fmoc-4-thi-oh, fmoc-3-4-thiazolyl-alanine, fmoc-, a-4-thiazolyl-ala-oh, fmoc-3-4-thiazolyl-l-alanine
7020858
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1,3-thiazol-4-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=N4)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 179°C
Quantity 5 g
Formula Weight 394.44
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(4-thiazolyl)-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'