missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-trans-4-hydroxy-L-prolinol, 96%

Brand:  Alfa Aesar™ H51050.03

Additional Details : CAS : 61478-26-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417937

  • 128.00 € / 1g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

61478-26-0
217.265
UFJNFQNQLMGUTQ-JGVFFNPUSA-N
11275979
CC(C)(C)OC(=O)N1CC(CC1CO)O
C10H19NO4
MFCD02094386
n-boc-trans-4-hydroxy-l-prolinol, boc-hyp-ol, 2s,4r-tert-butyl 4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s,4r-4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, boc-trans-4-hydroxy-l-prolinol, 2s,4r-1-tert-butoxycarbonyl-4-hydroxy-2-hydroxymethyl pyrrolidine, 2s,4r-1-boc-4-hydroxy-2-hydroxymethyl pyrrolidine, n-boc-2s,4r-4-hydroxy-2-hydroxymethylpyrrolidine, 2s,4r-tert-butyl4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, trans-4-hydroxy-l-prolinol, n-boc protected
tert-butyl (2S,4R)-4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Specifications

N-Boc-trans-4-hydroxy-L-prolinol
61478-26-0
MFCD02094386
UFJNFQNQLMGUTQ-JGVFFNPUSA-N
tert-butyl (2S,4R)-4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
11275979
96%
1g
C10H19NO4
n-boc-trans-4-hydroxy-l-prolinol, boc-hyp-ol, 2s,4r-tert-butyl 4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s,4r-4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, boc-trans-4-hydroxy-l-prolinol, 2s,4r-1-tert-butoxycarbonyl-4-hydroxy-2-hydroxymethyl pyrrolidine, 2s,4r-1-boc-4-hydroxy-2-hydroxymethyl pyrrolidine, n-boc-2s,4r-4-hydroxy-2-hydroxymethylpyrrolidine, 2s,4r-tert-butyl4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, trans-4-hydroxy-l-prolinol, n-boc protected
CC(C)(C)OC(=O)N1CC(CC1CO)O
217.265
217.26
Videos
SDS
Documents
Certificates